Learn More
Cayman Chemical AzelastInhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 2087310 MG
Encompass_Preferred
A histamine H1 receptor antagonist (Ki = 1.26 nM); selective for histamine H1 over histamine H3 receptors (Ki = 158.49 nM), as well as α1A- and α1B-adrenergic receptors (Ki = 50.12 nM for both); reduces nose rubbing, sneezing, and nasal mucosa expression of IL-4 in a dust mite-induced mouse model of allergic rhinitis at 0.14 UG/animal; reduces infection by a lentivirus pseudotyped with SARS-CoV-2 spike glycoprotein in HEK293 cells expressing ACE2 at 7 UG/ml
Catalog No. 50-298-1689