Learn More
Medchemexpress LLC Azenosertib (ZN-c3) | 2376146-48-2 | 100.0% | C29H34N8O2 | 10MG

Supplier: Medchemexpress LLC HY13229510MG
Azenosertib (ZN-c3) is a selective, orally active Wee1 kinase inhibitor used in preclinical cancer research. It potently inhibits Wee1 (IC50 = 3.9 nM) and has demonstrated antitumor activity. The compound is supplied as a light-yellow solid with high reported purity and includes recommended solubility and storage information for both in vitro and in vivo applications.
- Selective Wee1 kinase inhibitor with IC50 of 3.9 nM.
- High reported purity (99.96%).
- Available as solid and pre-made DMSO solution formats for convenience.
- Soluble in DMSO and compatible with common in vivo formulations.
- Recommended storage conditions provided for powder and solvent aliquots.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.