Learn More
Newcomer Supply AZF (Acetic Zinc Formalin) Fixative, 1 Liter. Excellent replacement for Mercury based fixatives
SDP
Supplier:  Newcomer Supply 1009A
Encompass
Use: Excellent replacement for Mercury based fixative. Working Solution. MERCURY FREE! Newcomer Supply AZF (Acetic Zinc Formalin) Fixative is a ready-to-use fixative recommended for bone marrow clots and cores, lymph nodes, endoscopic biopsies and immunohistochemical (IHC) studies. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.
Catalog No. NC9998836