Learn More
Medchemexpress LLC Azide-PEG-amine (MW 2000) | 95.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140661100MG
Encompass_Preferred
Azide-PEG-amine (MW 2000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent.
  • Contains an azide group.
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules.
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
Catalog No. 50-003-54204