Learn More
Medchemexpress LLC Azide-PEG12-alcohol | 1821464-55-4 | MFCD13184946 | 99.8% | 571.66 g/mol | C24H49N3O12 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1408035MG
Encompass_Preferred
Azide-PEG12-alcohol is an azide-functionalized polyethylene glycol (PEG12) linker with a terminal hydroxyl group, formulated for click chemistry and bioconjugation workflows. It enables efficient azide-alkyne cycloaddition and strain-promoted conjugation reactions and is commonly used in linker installation for degraders and conjugates.
  • Azide-functionalized PEG12 linker for bioconjugation.
  • Terminal hydroxyl group for further modification.
  • High purity (99.8%) for research applications.
  • Molecular weight approximately 571.66 g/mol.
  • Colorless to light yellow liquid; suitable for storage at recommended temperatures.
  • Compatible with CuAAC and SPAAC click reactions.
Catalog No. 50-002-22760