Learn More
Medchemexpress LLC Azide-PEG2-Ms | 176520-23-3 | ≥97.0% | C5H11N3O4S | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140381
Encompass_Preferred
Azide-PEG2-Ms is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that facilitates copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules possessing Alkyne groups. Additionally, it can engage in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker.
  • Can be used in the synthesis of PROTACs.
  • Click chemistry reagent.
  • Contains an azide group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with alkyne groups.
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
Catalog No. 50-003-52645