Learn More
Medchemexpress LLC Azide-PEG4-Tos | 168640-82-2 | 99.6% | C15H23N3O6S | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140351
Encompass_Preferred
Azide-PEG4-Tos is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This compound functions as a click chemistry reagent, enabling various bioconjugation strategies.
  • Contains an azide group for click chemistry reactions.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne groups.
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • Useful for the synthesis of PROTACs.
Catalog No. 50-003-52861