Learn More
Medchemexpress LLC Azide-PEG4-VC-PAB-doxorubicin | 99.6% | 1222.25 | C57H75N9O21 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1362881MG
Encompass_Preferred
Azide-PEG4-VC-PAB-doxorubicin is a drug-linker conjugate of doxorubicin with an azide-terminated PEG4-VC-PAB linker, intended for use in antibody-drug conjugate (ADC) research and for bioorthogonal conjugation via azide-alkyne cycloaddition.
  • Enables click chemistry conjugation via CuAAC or SPAAC reactions
  • Suitable for synthesis of antibody-agent conjugates (ADCs)
  • Provided as an orange to red solid and as a 10 mM solution in DMSO
  • High purity for research applications
  • Stable under recommended frozen and refrigerated storage conditions
Catalog No. 50-002-02711