Learn More
Medchemexpress LLC Azide-PEG4-VC-PAB-doxorubicin | 99.6% | 1222.25 | C57H75N9O21 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY13628825MG
Encompass_Preferred
Azide-PEG4-VC-PAB-doxorubicin is an azide-functionalized PEG4 val-cit-PAB linker conjugated to doxorubicin used to prepare antibody-drug conjugates and for click-chemistry applications.
  • Contains an azide group for copper-catalyzed and strain-promoted azide-alkyne cycloaddition reactions.
  • Includes a protease-cleavable Val-Cit-PAB linker for intracellular payload release.
  • Contains doxorubicin as the cytotoxic payload (anthracycline class).
  • High listed purity suitable for research (soluble in DMSO at 100 mg/mL).
  • Store as a solid at recommended low temperatures for long-term stability.
Catalog No. 50-002-02597