Learn More
Medchemexpress LLC Azide-PEG5-Tos | 236754-49-7 | MFCD28505520 | 98.5% | 417.48 g/mol | C17H27N3O7S | 250 MG

Supplier: Medchemexpress LLC HY140352250MG
Azide-PEG5-Tos is a cleavable five-unit polyethylene glycol (PEG5) linker bearing an azide functional group and a tosyl leaving group. It is used as a click-chemistry compatible linker and crosslinker for antibody-drug conjugates (ADCs) and other bioconjugation applications.
- Cleavable five-unit PEG linker suitable for ADC synthesis.
- Azide functional group enables CuAAC and strain-promoted cycloaddition.
- High purity suitable for research applications.
- Soluble in DMSO at ≥100 mg/mL.
- Molecular weight 417.48 g/mol; chemical formula C17H27N3O7S.
- Available in milligram to gram pack sizes, including 250 MG.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.