Learn More
Medchemexpress LLC Azido-methyltetrazine di-arm linker | 2861998-16-3 | 440.50 g·mol⁻¹ | C20H28N10O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY159794100MG
Encompass_Preferred
Azido-methyltetrazine di-arm linker is a bifunctional small-molecule linker designed for constructing multivalent antibody-drug conjugates (ADCs). It provides an azide handle and a methyltetrazine moiety to enable orthogonal bioorthogonal conjugation strategies for site-selective payload attachment in chemical biology and ADC development.
  • Bifunctional linker with azide and methyltetrazine groups.
  • Enables rapid inverse-electron-demand Diels-Alder ligation.
  • Compatible with click chemistry for selective payload attachment.
  • Supplied as a solid in multiple small-scale quantities for research use.
  • Molecular formula C20H28N10O2; molecular weight 440.5 g·mol⁻¹.
Catalog No. 50-002-02521