Learn More
Medchemexpress LLC Azido-methyltetrazine di-arm linker | 2861998-16-3 | 98.8% | 440.50 g/mol | C20H28N10O2 | 25 MG

Supplier: Medchemexpress LLC HY15979425MG
Azido-methyltetrazine di-arm linker is a bifunctional tetrazine-containing linker used for the synthesis of antibody-drug conjugates (ADCs) and other bioconjugates. It provides a di-arm scaffold that enables multivalent payload attachment via azide/tetrazine chemistries.
- Multivalent di-arm linker suitable for ADC synthesis.
- Purity 98.8% (analytical grade).
- Molecular weight 440.50 g/mol.
- Molecular formula C20H28N10O2.
- Appearance: red to purplish red solid.
- Available in small-scale packages, including 25 MG.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.