Learn More
Medchemexpress LLC Azido-methyltetrazine di-arm linker | 2861998-16-3 | 98.8% | 440.50 g/mol | C20H28N10O2 | 5 MG

Supplier: Medchemexpress LLC HY1597945MG
Azido-methyltetrazine di-arm linker is a multivalent linker used for the synthesis of antibody-drug conjugates (ADCs). It contains azide and methyltetrazine reactive groups to enable site-specific bioconjugation and click chemistry, and is supplied as a high-purity research reagent.
- Multivalent di-arm linker suitable for ADC construction.
- Contains azide and methyltetrazine functional groups for click chemistry.
- Molecular weight approximately 440.50 g/mol and formula C20H28N10O2.
- High purity suitable for research applications.
- Available in small research quantities for lab-scale conjugation experiments.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.