Learn More
Medchemexpress LLC Azido-methyltetrazine di-arm linker | 2861998-16-3 | 98.8% | 440.50 g/mol | C20H28N10O2 | 50 MG

Supplier: Medchemexpress LLC HY15979450MG
Azido-methyltetrazine di-arm linker is a multivalent antibody-drug conjugate (ADC) linker bearing azido and methyltetrazine functional groups to enable bioorthogonal conjugation during ADC synthesis. It is supplied as a red to purplish red solid with high purity for research use only.
- High purity (about 98.8%).
- Molecular weight 440.50 g/mol.
- Chemical formula C20H28N10O2.
- Red to purplish red solid appearance.
- Available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg, and 500 mg sizes.
- Storage: -20°C sealed; in solvent store at -80°C (6 months) or -20°C (1 month).
- Intended for research use only; not for human or animal therapeutic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.