Learn More
Medchemexpress LLC Azido-mono-amide-DOTA | 1227407-76-2 | 100.0% | 486.52 g·mol⁻¹ | C19H34N8O7 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW539916100MG
Encompass_Preferred
Azido-mono-amide-DOTA is a bifunctional macrocyclic DOTA derivative bearing an azide functional group. It is designed to chelate radiometals while providing an orthogonal handle for bioorthogonal conjugation of peptides and targeting ligands, enabling radiolabeling and tumor pre-targeting workflows.
  • High purity (≈99.96%).
  • Water soluble (≥100 mg/mL).
  • Azide functional group facilitates click chemistry conjugation.
  • Macrocyclic DOTA core provides robust radiometal chelation.
  • Suitable for peptide conjugation and radiolabeling applications.
Catalog No. 50-002-84095