Learn More
Medchemexpress LLC Azido-PEG(4)-Val-Cit-PAB-PNP | 1869126-60-2 | C₃₆H₅₁N₉O₁₃ | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY151726
Encompass_Preferred
Azido-PEG(4)-Val-Cit-PAB-PNP is a cleavable peptide Antibody-Drug Conjugate (ADC) linker. It functions as a click chemistry reagent, containing an azide group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules, and ring strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
  • Cleavable peptide ADC linker
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Enables ring strain-promoted alkyne-azide cycloaddition (SPAAC)
Catalog No. 50-003-74533