Learn More
Medchemexpress LLC Azido-PEG(4)-Val-Cit-PAB-PNP | 1869126-60-2 | C36H51N9O13 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY151726100MG
Encompass_Preferred
Azido-PEG(4)-Val-Cit-PAB-PNP is a cleavable peptide ADC linker and a click chemistry reagent containing an azide group.
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups.
  • Can participate in ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
Catalog No. 50-003-74204