Learn More
Nanocs Azido PEG Maleimide, N3-PEG-Ma 100mg. Azide/azido functionalized polyethylene glycol (N3-PEG-X) is a bifunctional PEG derivative that can be used to modify proteins, peptides and other materials. Azide group can react with alkyne in aqueous soluti
Supplier:  Nanocs PG2AZML600
Encompass
Azido PEG Maleimide, N3-PEG-Ma 100 mg
Catalog No. NC2858421