Learn More
Medchemexpress LLC Azido-PEG1 | 1517-05-1 | 100.0% | 87.08 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY138461
Encompass_Preferred
Azido-PEG1 is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an azide group.
  • Enables copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules that have alkyne groups.
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
Catalog No. 50-003-51566