Learn More
Medchemexpress LLC Azido-PEG1-amine | 464190-91-8 | ≥98.0% | C4H10N4O | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140212100MG
Encompass_Preferred
Azido-PEG1-amine is a PEG-based PROTAC linker designed for the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups. PROTACs utilize two ligands connected by a linker to target an E3 ubiquitin ligase and a protein of interest, facilitating selective degradation via the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Functions as a click chemistry reagent
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Utilized in PROTAC synthesis
  • Relevant for cancer targeted therapy
Catalog No. 50-003-57112