Learn More
Medchemexpress LLC Azido-PEG1-Boc | 1374658-85-1 | 98.0% | C9H17N3O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14077550MG
Encompass_Preferred
Azido-PEG1-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is a click chemistry reagent containing an Azide group. It can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups, and also strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Used in the synthesis of PROTACs
Catalog No. 50-003-52963