Learn More
Medchemexpress LLC Azido-PEG1-C2-acid | 1393330-34-1 | ≥98.0% | 159.14 | 25MG
SDP
Supplier:  Medchemexpress LLC 25MGHY140009
Encompass_Preferred
Azido-PEG1-C2-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that allows it to participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with Alkyne-containing molecules. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-50146