Learn More
Medchemexpress LLC Azido-PEG1-CH2CO2H | 79598-48-4 | 99.58% | 145.12 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY108369100MG
Encompass_Preferred
Azido-PEG1-CH2CO2H is a PROTAC linker, characterized by its alkyl/ether composition. This reagent is utilized in the synthesis of PROTAC BRD4 Degrader-1. It functions as a click chemistry reagent, containing an Azide group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with Alkyne group-containing molecules. Additionally, it can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • Utilized in the synthesis of PROTAC BRD4 Degrader-1
  • Functions as a click chemistry reagent
  • Contains an azide group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-46873