Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azido-PEG1-CH2CO2H | 79598-48-4 | 99.6% | 145.12 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY10836950MG

Encompass_Preferred

Azido-PEG1-CH2CO2H is a PROTAC linker with an alkyl/ether composition. It is utilized in the synthesis of PROTAC BRD4 Degrader-1. This compound serves as a click chemistry reagent, featuring an Azide group, which allows it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. Additionally, it can engage in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. It is intended for research use only.

  • Purity of 99.58%
  • Molecular weight of 145.12
  • Molecular formula C4H7N3O3
  • CAS number 79598-48-4
  • Appears as a colorless to light yellow liquid
  • Ships at room temperature
  • Store pure form at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at concentrations of 50 mg/mL or more

Catalog No. 50-003-47169


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.