Learn More
Medchemexpress LLC Azido-PEG10-amine | 912849-73-1 | ≥97.0% | C22H46N4O10 | 1 G

Supplier: Medchemexpress LLC HY1402171G
Azido-PEG10-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules possessing Alkyne groups. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. PROTACs consist of two different ligands connected by a linker, leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
- PEG-based PROTAC linker
- Used in the synthesis of PROTACs
- Click chemistry reagent
- Contains an Azide group for CuAAc and SPAAC reactions
- Appearance is colorless to off-white liquid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.