Learn More
Medchemexpress LLC Azido-PEG11-azide | 1392284-57-9 | 98.0% | C24H48N6O11 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13339610MG
Encompass_Preferred
Azido-PEG11-azide is a polyethylene glycol (PEG)-based bifunctional linker terminated with azide groups at both ends. It is supplied as a high-purity reagent for bioconjugation and PROTAC synthesis and is suitable for both copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC) reactions.
  • Terminal azide groups for efficient click chemistry conjugation.
  • PEG11 spacer provides flexibility and aqueous solubility.
  • High purity supports reproducible synthetic and biological studies.
  • Compatible with CuAAC and SPAAC reaction conditions.
  • Useful as a linker in PROTAC and bioconjugation applications.
Catalog No. 50-002-60375