Learn More
Medchemexpress LLC Azido-PEG12-acid | 2152679-71-3 | 99.8% | C27H53N3O14 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140456
Encompass_Preferred
Azido-PEG12-acid is a PEG-based PROTAC linker that functions as a click chemistry reagent. It contains an Azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. PROTACs utilize the intracellular ubiquitin-proteasome system to degrade target proteins.
  • Used in the synthesis of PROTACs
  • Click chemistry reagent with Azide group
  • Participates in CuAAc reactions
  • Participates in SPAAC reactions
  • Utilizes the ubiquitin-proteasome system for protein degradation
Catalog No. 50-003-52724