Learn More
Medchemexpress LLC Azido-PEG12-acid | 2152679-71-3 | 99.8% | C27H53N3O14 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14045650MG
Encompass_Preferred
Azido-PEG12-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent containing an Azide group, enabling both copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Alkyne groups and strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups. PROTACs leverage the ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent with Azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC)
  • PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-56927