Learn More
Medchemexpress LLC Azido-PEG2-C1-Boc | 251564-45-1 | C10H19N3O4 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140793
Encompass_Preferred
Azido-PEG2-C1-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is a versatile click chemistry reagent.
  • Click chemistry reagent containing an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups
Catalog No. 50-003-52740