Learn More
Medchemexpress LLC Azido-PEG2-C2-acid | 1312309-63-9 | 99.9% | C7H13N3O4 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1404521G
Encompass_Preferred
Azido-PEG2-C2-acid is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. This compound functions as a click chemistry reagent, featuring an azide group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing alkyne groups. Additionally, it can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Functions as a click chemistry reagent
  • Features an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Reacts with molecules containing alkyne, DBCO, or BCN groups
  • Utilizes intracellular ubiquitin-proteasome system for protein degradation
Catalog No. 50-003-53222