Learn More
Medchemexpress LLC Azido-PEG2-C2-amine | 166388-57-4 | MFCD13189985 | >97.0% | 174.20 g·mol⁻¹ | C6H14N4O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140213100MG
Encompass_Preferred
Azido-PEG2-C2-amine is a PEG-based bifunctional linker bearing an azide and a primary amine. It is used as a click chemistry reagent for copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC), and as a linker in PROTAC and antibody-drug conjugate (ADC) synthesis.
  • Azide functional group for click chemistry (CuAAC and SPAAC).
  • Primary amine for conjugation to carboxyl and activated ester groups.
  • PEG2 spacer provides water solubility and flexible linker length.
  • Typically supplied as a solid suitable for standard conjugation workflows.
  • Reported purity >97.0% and molecular weight 174.20 g·mol⁻¹.
Catalog No. 50-002-02625