Learn More
Medchemexpress LLC Azido-PEG2-C6-Cl | 2568146-55-2 | 95.0% | 249.74 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13847050MG
Encompass_Preferred
Azido-PEG2-C6-Cl is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, allowing it to participate in various cycloaddition reactions. PROTACs leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • Functions as a click chemistry reagent
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Utilized in the synthesis of PROTACs
Catalog No. 50-003-51015