Learn More
Medchemexpress LLC Azido-PEG2-hydrazide-Boc | 2100306-56-5 | 97.0% | C12H23N5O5 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140816
Encompass_Preferred
Azido-PEG2-hydrazide-Boc is a PEG-based PROTAC linker. It is used in the synthesis of PROTACs and is a click chemistry reagent. This compound contains an Azide group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Click chemistry reagent with azide group
  • Capable of copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-52763