Learn More
Medchemexpress LLC Azido-peg2-hydrazide-boc | 2100306-56-5 | MFCD30828695 | 95% | C12H23N5O5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140816100MG
Encompass_Preferred
Azido-PEG2-hydrazide-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. It acts as a click chemistry reagent, featuring an azide group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • Utilized in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-52983