Learn More
Medchemexpress LLC Azido-PEG23-C2-azide | 95.0% | 1125.30 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY138358
Encompass_Preferred
Azido-PEG23-C2-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is a click chemistry reagent containing an Azide group.
  • Can participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with Alkyne groups
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-48544