Learn More
Medchemexpress LLC Azido-PEG23-C2-azide | 95.0% | 1125.30 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13835850MG
Encompass_Preferred
Azido-PEG23-C2-azide is a PEG-based PROTAC linker designed for synthesizing PROTACs. This click chemistry reagent contains an azide group, enabling its use in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules, or strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN group-containing molecules.
  • PEG-based PROTAC linker
  • Used for PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an azide group for reactivity
  • Compatible with copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Compatible with strain-promoted alkyne-azide cycloaddition (SPAAC)
Catalog No. 50-003-51122