Learn More
Medchemexpress LLC Azido-PEG24-acid | 2195075-62-6 | 99.9% | C51H101N3O26 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY140457
Encompass_Preferred
Azido-PEG24-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This click chemistry reagent features an Azide group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne-containing molecules. It also participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with compounds containing DBCO or BCN groups.
  • Peg-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Click chemistry reagent
  • Features an Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-55934