Learn More
Medchemexpress LLC Azido-PEG24-acid | 2195075-62-6 | 99.9% | C51H101N3O26 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14045750MG
Encompass_Preferred
Azido-PEG24-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, allowing it to participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne-containing molecules, and also in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Functions as a click chemistry reagent.
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc).
  • Engages in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.
  • Store stock solution at -80°C for up to 6 months.
  • Store stock solution at -20°C for up to 1 month.
  • Requires sealed storage, away from moisture.
Catalog No. 50-003-56659