Learn More
Medchemexpress LLC Azido-PEG3-C6-Cl | 1625717-44-3 | 95.30% | 293.79 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY138466
Encompass_Preferred
Azido-PEG3-C6-Cl is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. It functions as a click chemistry reagent, capable of undergoing both copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne groups and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Click chemistry reagent
  • Contains an azide group
  • Participates in CuAAc reactions with alkyne groups
  • Undergoes SPAAC reactions with DBCO or BCN groups
Catalog No. 50-003-50480