Learn More
Broadpharm Azido-PEG3-NHS ester | 1245718-89-1 | MFCD23726613 | 500 mg
Supplier:  Broadpharm BP21605500MG
Encompass
Azido-PEG3-NHS ester | Purity: 98% | Mol Wt: 344.3 | 1245718-89-1 | MFCD23726613 | 500 mg
Catalog No. NC1171837