Learn More
Medchemexpress LLC Azido-PEG4-alcohol | 86770-67-4 | 98.9% | 219.24 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY22340
Encompass_Preferred
Azido-PEG4-alcohol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Azide group capable of undergoing copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules possessing Alkyne groups. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
Catalog No. 50-004-02884