Learn More
Medchemexpress LLC Azido-PEG4-Amido-tri-(t-butoxycarbonylethoxymethyl)-methane | 1421933-29-0 | 97.0% | C36H66N4O14 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140874100MG
Encompass_Preferred
Azido-PEG4-Amido-tri-(t-butoxycarbonylethoxymethyl)-methane is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group. This enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with Alkyne-containing molecules and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with compounds containing DBCO or BCN groups. PROTACs leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Click chemistry reagent
  • Features an Azide group
  • Enables CuAAc and SPAAC reactions
  • Supports PROTACs in degrading target proteins
Catalog No. 50-003-54567