Learn More
Medchemexpress LLC Azido-PEG4-Amido-Tris | 1398044-55-7 | 96.4% | C15H30N4O8 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14124950MG
Encompass_Preferred
Azido-PEG4-Amido-Tris is a PEG-based PROTAC linker used in the synthesis of PROTACs. This compound functions as a click chemistry reagent, featuring an azide group. It can engage in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing alkyne groups, and can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent with an azide group
  • Capable of copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-56027