Learn More
Medchemexpress LLC Azido-PEG4-azide | 182760-73-2 | ≥97.0% | 288.30 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY138692500MG
Encompass_Preferred
Azido-PEG4-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a click chemistry reagent containing an azide group. This allows it to undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-50515