Learn More
Medchemexpress LLC Azido-PEG4-(CH2)3-methyl ester | 1835759-71-1 | C13H25N3O6 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14085450MG
Encompass_Preferred
Azido-PEG4-(CH2)3-methyl ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an azide group. This allows it to participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules possessing alkyne groups. It can also engage in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker for PROTAC synthesis.
  • Click chemistry reagent.
  • Contains an azide group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with alkyne groups.
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
Catalog No. 50-003-55158