Learn More
Medchemexpress LLC Azido-PEG4-CH2-Boc | 864681-04-9 | ≥97.0% | 333.38 g/mol | C14H27N3O6 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY42618500MG
Encompass_Preferred
Azido-PEG4-CH2-Boc is a Boc-protected, azide-functionalized PEG4 linker designed for bioconjugation and linker synthesis in research applications. It provides an azide handle for click chemistry and a Boc-protected amine for downstream modification.
  • Azide functionality compatible with copper-catalyzed and strain-promoted click reactions.
  • Boc-protected amine handle for controlled deprotection and conjugation.
  • Four-unit PEG spacer for improved solubility and flexibility in conjugates.
  • Suitable for ADC and PROTAC linker development and general bioconjugation workflows.
  • High purity and defined molecular weight support reproducible synthetic results.
Catalog No. 50-002-02325