Learn More
Medchemexpress LLC Azido-PEG4-hydrazide | 2170240-96-5 | 99.6% | C11H23N5O5 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY140814
Encompass_Preferred
Azido-PEG4-hydrazide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an azide group. It can participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc)
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Exploits the intracellular ubiquitin-proteasome system
Catalog No. 50-003-52634