Learn More
Medchemexpress LLC Azido-PEG4-hydrazide | 2170240-96-5 | 99.6% | C11H23N5O5 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140817
Encompass_Preferred
Azido-PEG4-hydrazide is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an azide group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules featuring DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-52764