Learn More
Medchemexpress LLC Azido-PEG4-hydrazide-Boc | 1919045-01-4 | >98.0% | C16H31N5O7 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140817100MG
Encompass_Preferred
Azido-PEG4-hydrazide-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent containing an Azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules having Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. PROTACs exploit the intracellular ubiquitin-proteasome system to degrade target proteins, consisting of two different ligands connected by a linker: one for an E3 ubiquitin ligase and one for the target protein.
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-53162