Learn More
Medchemexpress LLC Azido-PEG5-acid | 1425973-16-5 | >98.0% | 335.35 | C13H25N3O7 | 10 G
SDP
Supplier:  Medchemexpress LLC HY13057210G
Encompass_Preferred
Azido-PEG5-acid is an azide-terminated five-unit polyethylene glycol (PEG5) acid linker used for bioconjugation and linker synthesis. It functions as a click-chemistry reagent capable of undergoing azide-alkyne cycloaddition reactions and is commonly used in PROTAC and antibody-drug conjugate assembly.
  • Azide functional group enables copper-catalyzed and strain-promoted click reactions.
  • Five-unit PEG spacer provides solubility and flexibility for conjugation.
  • High purity supports synthetic and bioconjugation applications.
  • Available in multiple pack sizes for scalable workflows.
  • Stable under recommended storage conditions to preserve reactivity.
Catalog No. 50-002-04030